Assay Details
| Gene Symbol: Gja1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012567.2 | NP_036699.1 | 2 | 3040 | 111 | |
| XM_006256503.2 | XP_006256565.1 | 2 | 2923 | 111 | ||
| GenBank mRNA | AY324140.1 | - | 2 | 3040 | 111 | |
| BC081842.1 | - | 2 | 3038 | 7105924 | 111 | |
More Information
Set Membership: |
|

Gja1
Genomic Map