Assay Details
| Gene Symbol: Mapre1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138509.3 | NP_612518.2 | 7 | 1979 | 157 | |
| XM_006235294.2 | XP_006235356.1 | 7 | 1962 | 157 | ||
| XM_006235295.2 | XP_006235357.1 | 7 | 2032 | 157 | ||
| GenBank mRNA | BC081726.1 | - | 7 | 1979 | 7192469 | 157 |
| FQ220232.1 | - | 1 | 586 | 157 | ||
| FQ226262.1 | - | 1 | 991 | 157 | ||
| FQ231004.1 | - | 1 | 918 | 157 | ||
| FQ233697.1 | - | 1 | 989 | 157 | ||
More Information
Set Membership: |
|

Mapre1
Genomic Map