Assay Details
| Gene Symbol: Zscan18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228133.2 | XP_006228195.1 | 2 | 961 | 90 | |
| XM_006228135.2 | XP_006228197.1 | 2 | 324 | 90 | ||
| XM_006228136.2 | XP_006228198.1 | 2 | 325 | 90 | ||
| XM_008758843.1 | XP_008757065.1 | 2 | 323 | 90 | ||
| XM_008758844.1 | XP_008757066.1 | 1 | 152 | 90 | ||
More Information
Set Membership: |
|

Zscan18
Genomic Map