Assay Details
| Gene Symbol: RGD1561430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228847.2 | XP_006228909.1 | 1 | 758 | 86 | |
| XM_008759258.1 | XP_008757480.1 | 1 | 758 | 86 | ||
More Information
Set Membership: |
|

RGD1561430
Genomic Map