Assay Details
| Gene Symbol: Fam43b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239294.2 | XP_006239356.1 | 1 | 1609 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006239294.2 transcripts.
Set Membership: |
|

Fam43b
Genomic Map