Assay Details
| Gene Symbol: Vimp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173120.2 | NP_775143.2 | 6 | 634 | 82 | |
| GenBank mRNA | AF367467.1 | - | 6 | 644 | 82 | |
| BC059111.1 | - | 6 | 638 | 6920258 | 82 | |
| FQ210095.1 | - | 6 | 626 | 82 | ||
| FQ218853.1 | - | 6 | 644 | 82 | ||
| FQ219944.1 | - | 6 | 626 | 82 | ||
| FQ220082.1 | - | 6 | 629 | 82 | ||
| FQ225575.1 | - | 6 | 645 | 82 | ||
| FQ229642.1 | - | 6 | 626 | 82 | ||
More Information
Set Membership: |
|

Vimp
Genomic Map