Assay Details
| Gene Symbol: Bles03 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024233.2 | NP_001019404.2 | 2 | 1454 | 57 | |
| NM_001243534.1 | NP_001230463.1 | 2 | 1468 | 57 | ||
| GenBank mRNA | AF502571.2 | - | 1 | 661 | 57 | |
| BC093387.1 | - | 2 | 1313 | 7389419 | 57 | |
| FQ233132.1 | - | 2 | 1347 | 57 | ||
More Information
Set Membership: |
|

Bles03
Genomic Map