Assay Details
| Gene Symbol: Amer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244725.2 | XP_006244787.1 | 3 | 4482 | 93 | |
| XM_576539.5 | XP_576539.3 | 2 | 4381 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Amer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244725.2 | XP_006244787.1 | 3 | 4482 | 93 | |
| XM_576539.5 | XP_576539.3 | 2 | 4381 | 93 | ||
Set Membership: |
|