Assay Details
| Gene Symbol: Scd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031841.1 | NP_114029.1 | 6 | 3177 | 77 | |
| GenBank mRNA | AB032243.1 | - | 6 | 3177 | 77 | |
| BC061737.1 | - | 6 | 3117 | 5599134 | 77 | |
More Information
Set Membership: |
|

Scd2
Genomic Map