Assay Details
| Gene Symbol: Mrps17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105923.1 | NP_001099393.1 | 3 | 984 | 76 | |
| XM_006249264.2 | XP_006249326.1 | 3 | 704 | 76 | ||
| GenBank mRNA | FQ223326.1 | - | 3 | 658 | 76 | |
More Information
Set Membership: |
|

Mrps17
Genomic Map