Assay Details
| Gene Symbol: LOC100910528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748683.3 | XP_003748731.1 | 2 | 455 | 82 | |
| XM_006227802.2 | XP_006227864.1 | 2 | 449 | 82 | ||
| XM_006227803.2 | XP_006227865.1 | 2 | 366 | 82 | ||
| XM_006227804.2 | XP_006227866.1 | 2 | 392 | 82 | ||
| Gene Symbol: Mrpl36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108879.1 | NP_001102349.1 | 2 | 439 | 82 | |
| XM_006227762.2 | XP_006227824.1 | 2 | 455 | 82 | ||
| XM_006227763.2 | XP_006227825.1 | 2 | 449 | 82 | ||
| XM_006227764.2 | XP_006227826.1 | 2 | 366 | 82 | ||
| XM_006227765.2 | XP_006227827.1 | 2 | 392 | 82 | ||
| GenBank mRNA | BC167015.1 | - | 2 | 386 | 9099683 | 82 |
More Information
Set Membership: |
|

LOC100910528
Genomic Map