Assay Details
| Gene Symbol: Nutf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007629.1 | NP_001007630.1 | 5 | 442 | 84 | |
| XM_006255480.2 | XP_006255542.1 | 4 | 406 | 84 | ||
| GenBank mRNA | BC061569.1 | - | 5 | 442 | 6886638 | 84 |
More Information
Set Membership: |
|

Nutf2
Genomic Map