Assay Details
| Gene Symbol: Zfp37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058209.2 | NP_478116.2 | 1338 | 72 | ||
| GenBank mRNA | AF072439.1 | - | 1369 | 72 | ||
| BC166844.1 | - | 1312 | 72 | |||
| Gene Symbol: LOC100363405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763746.1 | XP_008761968.1 | 1319 | 72 | ||
More Information
Set Membership: |
|

Zfp37
Genomic Map