Assay Details
| Gene Symbol: Chst14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109639.1 | NP_001103109.1 | 1 | 1220 | 84 | |
| GenBank mRNA | BC166542.1 | - | 1 | 1165 | 7193817 | 84 |
More Information
Set Membership: |
|

Chst14
Genomic Map