Assay Details
Gene Symbol: Scgb3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013180.1 | NP_001013198.1 | 2 - 3 | 302 | 56 | |
GenBank mRNA | AF436841.1 | - | 2 - 3 | 302 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
