Assay Details
| Gene Symbol: Pold4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013195.1 | NP_001013213.1 | 2 - 3 | 315 | 111 | |
| XM_006230841.2 | XP_006230903.1 | 2 - 3 | 1050 | 111 | ||
| GenBank mRNA | BC062014.1 | - | 2 - 3 | 315 | 5621348 | 111 |
More Information
Set Membership: |
|

Pold4
Genomic Map