Assay Details
| Gene Symbol: RGD1564319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126290.1 | NP_001119762.1 | 1 | 1218 | 105 | |
| GenBank mRNA | BC160905.1 | - | 1 | 1218 | 7455398 | 105 |
More Information
Set Membership: |
|

RGD1564319
Genomic Map