Assay Details
| Gene Symbol: Eif2s3y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167666.1 | NP_001161138.1 | 2 - 3 | 137 | 93 | |
| GenBank mRNA | BC158736.1 | - | 2 - 3 | 137 | 9098876 | 93 |
| FJ775732.1 | - | 1 - 2 | 78 | 93 | ||
More Information
Set Membership: |
|

Eif2s3y
Genomic Map