Assay Details
| Gene Symbol: Icam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012967.1 | NP_037099.1 | 4 - 5 | 954 | 95 | |
| GenBank mRNA | BC081837.1 | - | 4 - 5 | 966 | 7122397 | 95 |
| D00913.1 | - | 4 - 5 | 954 | 95 | ||
| FQ228241.1 | - | 4 - 5 | 974 | 95 | ||
More Information
Set Membership: |
|

Icam1
Genomic Map