Assay Details
| Gene Symbol: Cd38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013127.1 | NP_037259.1 | 7 - 8 | 853 | 97 | |
| XM_006251072.2 | XP_006251134.1 | 7 - 8 | 1107 | 97 | ||
| GenBank mRNA | D29646.1 | - | 7 - 8 | 853 | 97 | |
| D30795.1 | - | 7 - 8 | 848 | 97 | ||
| FQ212353.1 | - | 7 - 8 | 858 | 97 | ||
More Information
Set Membership: |
|

Cd38
Genomic Map