Assay Details
| Gene Symbol: Txnl4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013891.1 | NP_001013913.1 | 2 - 3 | 358 | 73 | |
| XM_006255579.2 | XP_006255641.1 | 3 - 4 | 468 | 73 | ||
| GenBank mRNA | BC089962.1 | - | 2 - 3 | 358 | 7314335 | 73 |
More Information
Set Membership: |
|

Txnl4b
Genomic Map