Assay Details
| Gene Symbol: Zc2hc1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014079.1 | NP_001014101.1 | 1 - 2 | 88 | 75 | |
| GenBank mRNA | BC078967.1 | - | 1 - 2 | 88 | 7111557 | 75 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC078967.1, NM_001014079.1 transcripts.
Set Membership: |
|

Zc2hc1c
Genomic Map