Assay Details
| Gene Symbol: Adtrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014144.1 | NP_001014166.1 | 3 - 4 | 501 | 97 | |
| XM_006253833.2 | XP_006253895.1 | 4 - 5 | 627 | 97 | ||
| GenBank mRNA | BC088287.1 | - | 3 - 4 | 501 | 7321485 | 97 |
More Information
Set Membership: |
|

Adtrp
Genomic Map