Assay Details
| Gene Symbol: Adtrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014144.1 | NP_001014166.1 | 5 - 6 | 775 | 78 | |
| XM_006253833.2 | XP_006253895.1 | 6 - 7 | 901 | 78 | ||
| GenBank mRNA | BC088287.1 | - | 5 - 6 | 775 | 7321485 | 78 |
More Information
Set Membership: |
|

Adtrp
Genomic Map