Assay Details
| Gene Symbol: Cxcl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017496.1 | NP_001017496.1 | 3 - 4 | 294 | 97 | |
| GenBank mRNA | BC088260.1 | - | 3 - 4 | 294 | 7375992 | 97 |
More Information
Set Membership: |
|

Cxcl13
Genomic Map