Assay Details
| Gene Symbol: Cckbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013165.2 | NP_037297.1 | 4 - 5 | 872 | 107 | |
| GenBank mRNA | M99418.1 | - | 4 - 5 | 963 | 107 | |
| X79208.1 | - | 3 - 4 | 578 | 107 | ||
More Information
Set Membership: |
|

Cckbr
Genomic Map