Assay Details
| Gene Symbol: Ces1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017004.1 | NP_058700.1 | 1 - 2 | 92 | 87 | |
| GenBank mRNA | BC088251.1 | - | 1 - 2 | 92 | 7371252 | 87 |
More Information
Set Membership: |
|

Ces1c
Genomic Map