Assay Details
| Gene Symbol: Ces1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017004.1 | NP_058700.1 | 2 - 3 | 299 | 80 | |
| XM_006255172.2 | XP_006255234.1 | 2 - 3 | 316 | 80 | ||
| GenBank mRNA | BC088251.1 | - | 2 - 3 | 299 | 7371252 | 80 |
| D00362.1 | - | 2 - 3 | 358 | 80 | ||
| M20629.1 | - | 2 - 3 | 232 | 80 | ||
| X78489.1 | - | 1 - 2 | 223 | 80 | ||
More Information
Set Membership: |
|

Ces1c
Genomic Map