Assay Details
| Gene Symbol: Ces1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017004.1 | NP_058700.1 | 3 - 4 | 444 | 118 | |
| XM_006255172.2 | XP_006255234.1 | 3 - 4 | 461 | 118 | ||
| GenBank mRNA | BC088251.1 | - | 3 - 4 | 444 | 7371252 | 118 |
More Information
Set Membership: |
|

Ces1c
Genomic Map