Assay Details
| Gene Symbol: Ces1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017004.1 | NP_058700.1 | 6 - 7 | 833 | 89 | |
| XM_006255172.2 | XP_006255234.1 | 6 - 7 | 850 | 89 | ||
| GenBank mRNA | BC088251.1 | - | 6 - 7 | 833 | 7371252 | 89 |
| D00362.1 | - | 6 - 7 | 892 | 89 | ||
| M20629.1 | - | 6 - 7 | 766 | 89 | ||
| X78489.1 | - | 5 - 6 | 757 | 89 | ||
More Information
Set Membership: |
|

Ces1c
Genomic Map