Assay Details
| Gene Symbol: Pla2g5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017174.1 | NP_058870.1 | 4 - 5 | 508 | 71 | |
| XM_006239139.2 | XP_006239201.1 | 5 - 6 | 1307 | 71 | ||
| XM_006239140.2 | XP_006239202.1 | 5 - 6 | 1886 | 71 | ||
| XM_006239141.2 | XP_006239203.1 | 4 - 5 | 430 | 71 | ||
| XM_008764209.1 | XP_008762431.1 | 6 - 7 | 1421 | 71 | ||
| XM_008764210.1 | XP_008762432.1 | 4 - 5 | 2528 | 71 | ||
| XM_008764211.1 | XP_008762433.1 | 3 - 4 | 543 | 71 | ||
| GenBank mRNA | AY651028.1 | - | 4 - 5 | 375 | 71 | |
| BC085745.1 | - | 4 - 5 | 533 | 7192819 | 71 | |
| U03763.1 | - | 4 - 5 | 508 | 71 | ||
More Information
Set Membership: |
|

Pla2g5
Genomic Map