Assay Details
| Gene Symbol: Ldhc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017266.2 | NP_058962.2 | 1 - 2 | 92 | 96 | |
| GenBank mRNA | BC078862.1 | - | 1 - 2 | 92 | 7131609 | 96 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC078862.1, NM_017266.2 transcripts.
Set Membership: |
|

Ldhc
Genomic Map