Assay Details
| Gene Symbol: Lin7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021851.1 | NP_068623.1 | 1 - 2 | 33 | 78 | |
| GenBank mRNA | AF090136.1 | - | 1 - 2 | 33 | 78 | |
| FQ212254.1 | - | 65 | 78 | |||
More Information
Set Membership: |
|

Lin7c
Genomic Map