Assay Details
| Gene Symbol: Cxcl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022214.1 | NP_071550.1 | 2 - 3 | 239 | 82 | |
| XM_008770053.1 | XP_008768275.1 | 9 - 10 | 1636 | 82 | ||
| XM_008770054.1 | XP_008768276.1 | 2 - 3 | 656 | 82 | ||
| XM_008770055.1 | XP_008768277.1 | 9 - 10 | 1633 | 82 | ||
| GenBank mRNA | U90448.1 | - | 2 - 3 | 239 | 82 | |
More Information
Set Membership: |
|

Cxcl6
Genomic Map