Assay Details
| Gene Symbol: Cdx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023963.1 | NP_076453.1 | 2 - 3 | 788 | 65 | |
| XM_008768999.1 | XP_008767221.1 | 2 - 3 | 679 | 65 | ||
| GenBank mRNA | AJ278466.1 | - | 2 - 3 | 788 | 65 | |
More Information
Set Membership: |
|

Cdx2
Genomic Map