Assay Details
| Gene Symbol: Nfkbib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030867.2 | NP_110494.2 | 2 - 3 | 545 | 64 | |
| GenBank mRNA | AF246634.1 | - | 2 - 3 | 368 | 64 | |
| BC085729.1 | - | 2 - 3 | 545 | 7114624 | 64 | |
| FQ228042.1 | - | 2 - 3 | 342 | 64 | ||
| FQ235103.1 | - | 2 - 3 | 490 | 64 | ||
More Information
Set Membership: |
|

Nfkbib
Genomic Map