Assay Details
| Gene Symbol: Rab3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031091.2 | NP_112353.1 | 1 - 2 | 109 | 85 | |
| XM_006238531.2 | XP_006238593.1 | 1 - 2 | 155 | 85 | ||
| GenBank mRNA | CB711467.1 | - | 1 - 2 | 120 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_031091.2, XM_006238531.2 transcripts.
Set Membership: |
|

Rab3b
Genomic Map