Assay Details
| Gene Symbol: Klk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031523.1 | NP_113711.1 | 3 - 4 | 534 | 102 | |
| GenBank mRNA | BC078784.1 | - | 3 - 4 | 516 | 7128287 | 102 |
| J00758.1 | - | 3 - 4 | 528 | 102 | ||
| M11563.1 | - | 3 - 4 | 534 | 102 | ||
| X03560.1 | - | 2 - 3 | 352 | 102 | ||
More Information
Set Membership: |
|

Klk1
Genomic Map