Assay Details
| Gene Symbol: Ccnh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052981.2 | NP_443213.2 | 8 - 9 | 1171 | 107 | |
| GenBank mRNA | AF154914.1 | - | 7 - 8 | 868 | 107 | |
| AJ276495.1 | - | 8 - 9 | 1171 | 107 | ||
| BC059109.1 | - | 7 - 8 | 974 | 6886953 | 107 | |
More Information
Set Membership: |
|

Ccnh
Genomic Map