Assay Details
| Gene Symbol: Bhlhe40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053328.1 | NP_445780.1 | 1 - 2 | 400 | 87 | |
| GenBank mRNA | AF009330.1 | - | 1 - 2 | 400 | 87 | |
More Information
Set Membership: |
|

Bhlhe40
Genomic Map