Assay Details
| Gene Symbol: Kif3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053486.1 | NP_445938.1 | 1 - 2 | 1622 | 60 | |
| GenBank mRNA | AF083330.1 | - | 1 - 2 | 1592 | 60 | |
| AJ223599.1 | - | 1 - 2 | 1622 | 60 | ||
More Information
Set Membership: |
|

Kif3c
Genomic Map