Assay Details
| Gene Symbol: Pom121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053622.1 | NP_446074.1 | 1 - 2 | 578 | 102 | |
| GenBank mRNA | Z21513.1 | - | 1 - 2 | 578 | 102 | |
More Information
Set Membership: |
|

Pom121
Genomic Map