Assay Details
| Gene Symbol: Pom121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053622.1 | NP_446074.1 | 7 - 8 | 1365 | 63 | |
| XM_008769111.1 | XP_008767333.1 | 7 - 8 | 1854 | 63 | ||
| GenBank mRNA | Z21513.1 | - | 7 - 8 | 1365 | 63 | |
More Information
Set Membership: |
|

Pom121
Genomic Map