Assay Details
| Gene Symbol: Csrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057144.1 | NP_476485.1 | 3 - 4 | 448 | 86 | |
| XM_006229237.2 | XP_006229299.1 | 4 - 5 | 552 | 86 | ||
| GenBank mRNA | X81193.1 | - | 3 - 4 | 448 | 86 | |
More Information
Set Membership: |
|

Csrp3
Genomic Map