Assay Details
| Gene Symbol: Tas1r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130818.1 | NP_570831.1 | 2 - 3 | 492 | 60 | |
| XM_008764334.1 | XP_008762556.1 | 3 - 4 | 1288 | 60 | ||
| GenBank mRNA | AF456324.1 | - | 2 - 3 | 492 | 60 | |
| AY032620.1 | - | 2 - 3 | 492 | 60 | ||
More Information
Set Membership: |
|

Tas1r3
Genomic Map