Assay Details
| Gene Symbol: Calcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138513.1 | NP_612522.1 | 1 - 2 | 97 | 99 | |
| XM_006230075.2 | XP_006230137.1 | 3 - 4 | 571 | 99 | ||
| XM_008759674.1 | XP_008757896.1 | 4 - 5 | 426 | 99 | ||
| GenBank mRNA | M11596.1 | - | 1 - 2 | 97 | 99 | |
More Information
Set Membership: |
|

Calcb
Genomic Map