Assay Details
| Gene Symbol: Prl5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138527.1 | NP_612536.1 | 1 - 2 | 76 | 95 | |
| GenBank mRNA | AB022883.1 | - | 1 - 2 | 76 | 95 | |
| FQ220484.1 | - | 1 - 2 | 84 | 95 | ||
| FQ230176.1 | - | 1 - 2 | 84 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB022883.1, NM_138527.1 transcripts.
Set Membership: |
|

Prl5a1
Genomic Map