Assay Details
| Gene Symbol: Prl5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138527.1 | NP_612536.1 | 3 - 4 | 360 | 90 | |
| GenBank mRNA | AB022883.1 | - | 3 - 4 | 360 | 90 | |
| AF226607.1 | - | 3 - 4 | 330 | 89 | ||
| FQ220484.1 | - | 3 - 4 | 368 | 89 | ||
| FQ230176.1 | - | 3 - 4 | 368 | 89 | ||
More Information
Set Membership: |
|

Prl5a1
Genomic Map