Assay Details
| Gene Symbol: Syt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138835.1 | NP_620190.1 | 6 - 7 | 1181 | 68 | |
| XM_008760071.1 | XP_008758293.1 | 6 - 7 | 1109 | 68 | ||
| XM_008760072.1 | XP_008758294.1 | 7 - 8 | 1155 | 68 | ||
| GenBank mRNA | U71294.1 | - | 6 - 7 | 1181 | 68 | |
More Information
Set Membership: |
|

Syt12
Genomic Map