Assay Details
Gene Symbol: Prr21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139184.1 | NP_631923.1 | 1 - 2 | 103 | 94 | |
XM_008763356.1 | XP_008761578.1 | 1 - 2 | 102 | 94 | ||
GenBank mRNA | L08134.1 | - | 1 - 2 | 103 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
